missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (ALB/398), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76991AF700
This item is not returnable.
View return policy
Description
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) samples. It is validated for ELISA
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Purified Bovine Serum Albumin (Uniprot: P02769) | |
| 0.1 mL | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| 280717 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| ALB/398 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction