missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (ALB/398), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76991H
This item is not returnable.
View return policy
Description
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) samples. It is validated for ELISA
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| HRP | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Purified Bovine Serum Albumin (Uniprot: P02769) | |
| 0.1 mL | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| 280717 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| ALB/398 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction