Learn More
Trajan Scientific and Medical BP BOND Q PLOT Columns
Manufactured for stability at high temperatures, low bleed and high inertness levels. Designed for the analysis of fixed gases, low molecular weight hydrocarbons, volatile polar compounds and reactive analytes. Engineered to prevent the stationary phase particles from shredding downstream.
Brand:  Trajan Scientific and Medical 0570535
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15740949
  • € 769.00 / Each
Estimated Shipment
01-12-2020
Add to basket
Description
Description
  • Highly stable column can withstand repeated water injections
  • Reduced need for particle trap due to minimal particle shredding
  • Broad application range - ideal for volatile solvent and hydrocarbon analysis
Specifications
Specifications
PLOT
100% divinylbenzene
-100°C to +300/320°C
0.32 mm
5 µm
300/320°C
50 m