missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical BP BOND U PLOT Columns

Manufactured for stability at high temperatures, low bleed and high inertness levels. Designed for the analysis of fixed gases, low molecular weight hydrocarbons, volatile polar compounds and reactive analytes. Engineered to prevent the stationary phase particles from shredding downstream.

Brand:  Trajan Scientific and Medical 0571137

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15750949

  • € 349.00 / Each


Estimated Shipment
27-11-2020
Add to basket

Description

Description

  • Maximum temperature extended from 190°C to 30°C
  • Bleed reduction provides lower detection limits and faster stabilization times
  • Ideal for applications such as trace hydrogen sulfide
  • Carbonyl sulfide and mercaptans in hydrocarbon streams
Specifications

Specifications

PLOT
Divinylbenzene Ethylene Glycol, Dimethyl Acrylate
-100°C to +300°C
0.32 mm
7 µm
300°C
10 m
Videos
SDS
Documents

For research use only