missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BP1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-481UV
This item is not returnable.
View return policy
Description
BP1 Polyclonal specifically detects BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| BP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8 | |
| Rabbit | |
| 0.1 mL | |
| 1748 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DLX4 | |
| A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProtNo. Q92988]. | |
| Primary | |
| This is specific for human BP1 (DLX4 variant 1). Berg PE, Kirolikar S. DLX4 (distal-less homeobox 4). Atlas Genet Cytogenet Oncol Haematol. January 2011. URL : http://AtlasGeneticsOncology.org/Genes/DLX4ID49827ch17q21.html | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction