missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BPIL3 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | BPIL3 |
| Synonym | bactericidal/permeability-increasing protein-like 3, BPI fold containing family B, member 6, BPI fold-containing family B member 6, LPLUNC6 |
| Gene Symbol | BPIFB6 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction