missing translation for 'onlineSavingsMsg'
Learn More

BQU57, MedChemExpress

Product Code. 16491607
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491607

Brand: Medchem Express HY12875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BQU57 shows selective inhibition for Ral relative to Ras or Rho and inhibit xenograft tumor growth similar to depletion of Ral by siRNA. The IC50 for BQU57 of 2.0 (mu)M in H2122 and 1.3 (mu)M in H358.

BQU57 shows selective inhibition for Ral relative to Ras or Rho and inhibit xenograft tumor growth similar to depletion of Ral by siRNA. The IC50 for BQU57 of 2.0 (mu)M in H2122 and 1.3 (mu)M in H358.

Specifications

Chemical Name or Material BQU57
Physical Form Solid
CAS 1637739-82-2
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H13F3N4O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 100 mg/mL (299.13 mM)
SMILES NC1=C(C#N)C(C(C(C)=NN2C)=C2O1)C3=CC=C(C(F)(F)F)C=C3
Molecular Weight (g/mol) 334.3
Formula Weight 334.3
Percent Purity 99.55%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.