Learn More
Brachyury Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-87598
 View more versions of this product
Product Code. 18797133
  • € 539.00 / 0.10mL
Description
Brachyury Polyclonal specifically detects Brachyury in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.