missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brachyury Antibody (TBXT/7853R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21002FR
This item is not returnable.
View return policy
Description
Brachyury Monoclonal antibody specifically detects Brachyury in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Brachyury | |
| Monoclonal | |
| DyLight 680 | |
| brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT | |
| Recombinant fragment (around aa 235-435) of human Brachyury protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 6862 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TBXT/7853R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction