missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Brachyury Recombinant Rabbit Monoclonal Antibody (JE44-11)

Description
Positive Control: SH-SY-5Y, Mouse E14.5 embryo tissue.
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
Specifications
Specifications
| Antigen | Brachyury |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JE44-11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | TBXT |
| Gene Accession No. | O15178 |
| Gene Alias | Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?