Learn More
Brady™ BMP51/BMP53/BMP41 Label Maker Cartridge: B-428 Metalized Polyester
Click to view available options
Dimensions (LxW):
1 x 0.5 in.
1.13 x 0.5 in.
2 x 1 in.
Unit Size:
Each
Description
- Available in various sizes
- For use with BMP41, BMP51, BMP53 printers
Specifications
Specifications
| Certifications/Compliance | Pass 2002/95/EC |
| Color | Black, Light Gray |
| Material | Metallized Polyester (B-428) |
| Product Type | Label Maker Cartridge |
| Quantity | 480/Pk. |
| Size (English) | 1 x 0.5 in. |
| Size (Metric) | 25.4 x 12.7 mm |
| Dimensions (LxW) | 1 x 0.5 in. |
| Width (English) | 1 in. |
| Width (Metric) | 25.4 mm |
| Show More |