Learn More
Brady™ Brady™Bondz™ B-483 Ultra Aggressive Polyester BMP71 Printer Label
Click to view available options
Dimensions (LxW):
0.5 x 1 in.
Unit Size:
Each
Description
- Glossy white polyester with permanent rubber based adhesive is thermal transfer printable
- Material is designed for high adhesion to textured metals and low surface energy plastics
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
- Service temperature of -40 to 248° F (-40 to 120° C)
Specifications
Specifications
| Certifications/Compliance | UL Recognised; CSA Accepted |
| Description | BMP71 Label Printer Labels |
| Adhesive Type | Permanent Rubber Based |
| Color | White |
| Finish | Gloss |
| Material | Polyester |
| Sterility | Non-sterile |
| For Use With (Equipment) | BMP™71 Label Printer |
| Labels per Row | 1 |
| Quantity | 500/Pk. |
| Show More |