Learn More
Brady™ Globally Harmonized System (GHS) Flammable Picto Labels
Product Code. 15640351
Click to view available options
Quantity:
39/Pk.
Material:
Vinyl
Height (English):
0.62 in.
Width (English):
0.6 in.
Unit Size:
Pack of 39
Product Code. 15640351
Brand: Brady™ 118821
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For flammable gases, liquids and solids; pyrophorics; self-reatives; self-heating
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
  • Legend: Flammable
  • Made of Vinyl or Coated paper
  • Available in three sizes
Specifications
Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Adhesive Type Permanent Acrylic
Color Black, Red, White
Finish Gloss
Format Label
Material Vinyl
Product Type Pictogram Label
Sterility Non-sterile
Display Text Flammable
For Use With (Application) Flammable gases, liquids and solids
Quantity 39/Pk.
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Dimensions (LxW) 0.625 x 0.625 in.
Width (English) 0.6 in.
Height (English) 0.62 in.
Height (Metric) 15.20 mm