Learn More
Brady™ LABXPERT™ Polyester Slide Labels
Product Code. p-3006902
Click to view available options
Height (Metric):
6.35 mm
Width (Metric):
4.82 cm
Unit Size:
1 roll
Product Code. 10123092
Brand: Brady™ 060952
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these labels for laboratory and healthcare applications. Brady™ LABXPERT™ Polyester Slide Labels are for use with LABXPERT™ and IDXPERT™ printers.
  • Withstand temperatures from -80°C to +121°C (-112°F to +250°F)
  • Material offers permanent adhesion, outstanding chemical resistance, and write-on capabilities
  • Resist xylene, toluene, DMSO, and ethanol
  • Specifications
    Certifications/Compliance AGA, UL, UL969, CSA, C22.2 No.0.15-95, RoHS, 2005/618/EC MCV, 2002/95/EC
    Description Surface Mount
    Adhesive Type Acrylic, Permanent
    Color Black, White
    Finish Matte
    Format Label
    Material Polyester
    Product Type Label
    For Use With (Application) Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates, Slide label
    For Use With (Equipment) iDXPERT™, LabXpert™ Printer
    Labels per Row 1
    Quantity 200/Pk.
    Shape Rectangle
    Temperature Range (Metric) -40°C to +160°C
    Thickness (Metric) 0.091 mm
    Width (English) 1.9 in.
    Width (Metric) 4.82 cm
    Height (English) 0.25 in.
    Height (Metric) 6.35 mm
    Height (Metric) Printable Area 6.35 mm