Learn More
Brady™ BradyGrip™ Print-on-Hook Labels featuring VELCRO™ Hook for M7 Printers
Product Code. 17889144
Click to view available options
Dimensions (LxW):
20 ft. x 0.25 in.
20 ft. x 0.5 in.
20 ft. x 0.75 in.
20 ft. x 1 in.
20 ft. x 1.5 in.
Unit Size:
Each
Product Code. 17889144
Brand: Brady™ 173238
For wire and cable ID, BradyGrip™'s Print-on Hook Material is a unique print-and-place material that can be easily applied, repositioned or removed.
  • Features a printable front and grippy back with tiny hooks
  • Attaches to VELCRO™ Brand ONE-WRAP™ Strap for easy wire and cable ID
  • Doesn't damage wires and cables like nylon cable ties
  • Maximizes your uptime with faster, printable ID on cable bundles
  • VELCRO™ Brand ONE-WRAP™ product not included with purchase
Specifications
Autoclavable Not Autoclavable
Adhesive Type Non-adhesive
Coating None
Color White
Finish Matte
Format Continuous
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Wire and Cable Labeling
For Use With (Equipment) M710, BMP71
Latex Free No
Quantity 20 ft.
Shape Rectangle
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.015 in.
Thickness (Metric) 0.381 mm
Dimensions (LxW) 20 ft. x 0.25 in.
Length (English) 20 ft.
Length (Metric) 6.1 m
Width (English) 0.25 in.
Width (Metric) 6.35 mm
Alternate Part Number M7C-250-414