missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bradykinin RB2/BDKRB2 Antibody (471916) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB47251JF635
This item is not returnable.
View return policy
Description
Bradykinin RB2/BDKRB2 Monoclonal antibody specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| Bradykinin RB2/BDKRB2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| B2 bradykinin receptor, B2R, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BRB2, DKFZp686O088 | |
| NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession # P30411 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 471916 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 624 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction