Learn More
BRAF Mouse anti-Human, Clone: OTI3E8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500441
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
B-raf belongs to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in the gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, maligt melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene.Specifications
| BRAF | |
| Monoclonal | |
| Unconjugated | |
| BRAF | |
| B-RAF1, B-raf, BRAF1, NS7, RAFB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 673 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15056 | |
| BRAF | |
| Full length human recombit protein of human BRAF produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.