missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAF Mouse anti-Human, Clone: OTI4G6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500445
This item is not returnable.
View return policy
Description
B-raf belongs to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in the gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, maligt melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene.Specifications
| BRAF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P15056 | |
| BRAF | |
| Full-length protein expressed in 293T cell transfected with human BRAF expression vector | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G6 | |
| Unconjugated | |
| BRAF | |
| B-RAF1, B-raf, BRAF1, NS7, RAFB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 673 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction