missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAF35 Antibody (PCRP-HMG20B-1B5), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20903DL594
This item is not returnable.
View return policy
Description
BRAF35 Monoclonal antibody specifically detects BRAF35 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| BRAF35 | |
| Monoclonal | |
| DyLight 594 | |
| BRAF35Sox-like transcriptional factor, high-mobility group 20B, HMG domain-containing protein 2, HMG domain-containing protein HMGX2, HMGX2FLJ26127, HMGXB2member 1-related, SMARCE1R, SMARCE1-related protein | |
| Synthetic peptide (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 10362 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PCRP-HMG20B-1B5 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction