missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Brain Basal Ganglia Tissue Slides (Adult Normal)- Paraffin
Shop All Bio Techne Products
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
Description
The Human Brain Basal Ganglia Tissue Lysate (Adult Normal) is derived from human.
Specifications
Specifications
| Quantity | 5 Slides |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Tissue Condition | Normal |
| Cross Reactivity | CD300d |
| Host Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Life Stage | Adult |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction