Learn More
BRAND™ BRANDplates™ Inlet Opening System PC Membrane Single Inserts
Product Code. 15600617 Shop All Brand Products
Click to view available options
Type:
PC Membrane 0.4 μm
PC Membrane 1.0 μm
PC Membrane 3.0 μm
PC Membrane 8.0 μm
Unit Size:
Pack of 48
Product Code. 15600617
Brand: BRAND™ 782807
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sterile, individual inserts
  • Innovative cell culture plates and inserts for manual and automated cell and tissue culture
  • The BRANDplates™ Insert System, developed jointly with the Fraunhofer Gesellschaft, forms an innovative extension of BRAND's existing cell culture range
  • It consists of two different cell culture plates and two versions of cell culture inserts, which can also be combined with each other
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description Inlet channels
    Type PC Membrane 0.4 μm
    For Use With (Equipment) 24-/6-well BRANDplates™
    Height (Metric) 13 mm
    Quantity 48