Learn More
BRAND™ Piston Heads for Titrette™ Bottletop Burette
Shop All Brand Products
Click to view available options
For Use With (Equipment):
10 mL Titrette
25 mL Titrette
Unit Size:
Each
Description
- Fits all serial number ranges
- Available for 10, 25 or 50mL Titrette™ Bottletop Burette
Specifications
Specifications
| Product Type | Piston Head |
| For Use With (Equipment) | 10 mL Titrette |