Learn More
BRAND™ BIO-CERT™ Filter Tips, 50 to 1000uL
Constructed of high-purity, oleamide- and diHEMDA-free polypropylene. BIO-CERT™ Filter Tips, 50 to 1000μL are sterile and free from biological contaminants. Non self-sealing PE filters allow sample recovery with wetted filters. Thin-walled for high translucency, easy ejection, and low waste.
Brand: BRAND™ 732734
127.19 EUR valid until 2024-05-31
Use promo code "21712" to get your promotional price.
Additional Details : Weight : 0.95490kg
Description
- High-purity polypropylene is oleamide- and diHEMDA-free
- Independently certified sterile and free from biological contaminants
- Non self-sealing PE filters allow sample recovery with wetted filters
- Thin-walled for high translucency, easy ejection, and low waste.
- Graduations at 250, 500, and 1000μL for quick volume check.
- Available in either standard or ultra low retention types
- Available in convenient, ecological rack format; racked tips are colorless and placed into blue-colored tip tray
Ultra low retention tips
- Produced through special, patented physicochemical process
- Homogeneous, defect-free surface for low surface tension and low sample retention
- Reduce sample loss and provide substantially higher reproducibility when working with critical media
- For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
- High chemical resistance
- Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties
Specifications
50 to 1000 μL | |
Sterile | |
Autoclavable | |
7cm | |
Rack | |
Bio-Cert | |
250 μL, 500 μL, 1000 μL |
Filtered | |
Colorless | |
2.75 in. | |
DNase, RNase, ATP, and Endotoxin Free | |
Universal | |
Polypropylene |