Learn More
BrandTech™ BRAND™ Transferpette™ Single-Channel Pipetters: Tapered Pipet Shaft Cone
Click to view available options
Functional Range:
2 to 20μL, 0.5 to 10μL
Unit Size:
Each
Description
- Does not fit Transferpette S models
Specifications
Specifications
| For Use With (Equipment) | 2 to 20μL digital ultra micro and 0.5 to 10μL Transferpette™ digital single channel pipette |
| Functional Range | 2 to 20μL, 0.5 to 10μL |
| Includes | Stainless Steel spring |