Learn More
BRANDTECH™ VITLAB™ Polypropylene Powder Funnels
Product Code. p-4380301
Change view
Click to view available options
Diameter (Metric) Stem:
28mm
Diameter (Metric)Top:
150mm
Height (Metric):
138 mm
Length (Metric) Stem:
22mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Stem Diameter (Metric)Top Height (Metric) Length (Metric) Stem unitSize
10007251 28mm 150mm 138 mm 22mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10007251 Supplier BRANDTECH™ VITLAB™ Supplier No. 71194
Add to basket
Add to basket
Available in a variety of sizes and stem diameters to fit your application.
  • Polypropylene
  • With volume markings
Specifications
Diameter (Metric)Top 150mm
Diameter (Metric) Stem 28mm
Material Polypropylene
Height (Metric) 138 mm
Length (Metric) Stem 22mm
Product Type Funnel