Learn More
BRANDTECH™ VITLAB™ Polypropylene Powder Funnels
Click to view available options
Diameter (Metric) Stem:
28mm
Diameter (Metric)Top:
150mm
Height (Metric):
138 mm
Length (Metric) Stem:
22mm
Unit Size:
Pack of 5
Description
- Polypropylene
- With volume markings
Specifications
Specifications
| Diameter (Metric)Top | 150mm |
| Diameter (Metric) Stem | 28mm |
| Material | Polypropylene |
| Height (Metric) | 138 mm |
| Length (Metric) Stem | 22mm |
| Product Type | Funnel |