Learn More
Branson Ultrasonics™ Sonifier Spanner Wrench
Product Code. 15747809
Click to view available options
For Use With (Equipment):
250 and 450 Sonifier Cell Disruptor
Unit Size:
Each
Product Code. 15747809
Brand: Branson Ultrasonics™ 101118039
Only null left
Add to basket
Add to basket
Requires
2 wrenches needed; one for converter and one for probe
Removes disruptor from 250 and 450 model Sonifiers™
Removes disruptor from 250 and 450 model Sonifiers™
Made from titanium
2 required; One for converter and one for probe
Specifications
Product Type Wrench Spanner
Material Titanium
For Use With (Equipment) 250 and 450 Sonifier Cell Disruptor