Learn More
Branson Ultrasonics™ Sonifier Spanner Wrench
Click to view available options
For Use With (Equipment):
250 and 450 Sonifier Cell Disruptor
Unit Size:
Each
Description
Requires
2 wrenches needed; one for converter and one for probe
Removes disruptor from 250 and 450 model Sonifiers™
Made from titanium
2 required; One for converter and one for probe
Specifications
Specifications
| Product Type | Wrench Spanner |
| Material | Titanium |
| For Use With (Equipment) | 250 and 450 Sonifier Cell Disruptor |