missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAP Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42696AF405
This item is not returnable.
View return policy
Description
BRAP Polyclonal antibody specifically detects BRAP in Human samples. It is validated for Western Blot
Specifications
| BRAP | |
| Polyclonal | |
| Western Blot | |
| BRAP2RING finger protein 52, BRCA1 associated protein, EC 6.3.2.-, IMPBRCA1-associated protein, Impedes mitogenic signal propagation, Renal carcinoma antigen NY-REN-63, RNF52galectin-2-binding protein | |
| The immunogen for this product maps to a region between residue 560 and 592 of human BRCA1 associated protein using the numbering given in entry NP_006759.3 (GeneID 8315). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8315 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction