missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Monoclonal Antibody (OTI7B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802819S
This item is not returnable.
View return policy
Description
BRCA1 Monoclonal antibody specifically detects BRCA1 in Human samples. It is validated for Western BlotSpecifications
| BRCA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P51398 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7818 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7B12 | |
| Unconjugated | |
| DAP3 | |
| brca 1; Brca1; brca-1; BRCA1, DNA repair associated; BRCA1/BRCA2-containing complex, subunit 1; BRCAI; BRCC1; breast and ovarian cancer susceptibility protein 1; breast and ovarian cancer sususceptibility protein 1; breast cancer 1; breast cancer 1 early onset; breast cancer 1, early onset; breast cancer associated 1; breast cancer type 1 susceptibility protein; Breast cancer type 1 susceptibility protein homolog; BROVCA1; BROVCA12; Delta772-3095; early onset breast cancer 1; Fanconi anemia, complementation group S; FANCS; IRIS; OTTHUMP00000212147; OTTHUMP00000212148; OTTHUMP00000212149; OTTHUMP00000212150; OTTHUMP00000212151; OTTHUMP00000212155; PNCA4; PPP1R53; protein phosphatase 1, regulatory subunit 53; PSCP; RING finger protein 53; RING-type E3 ubiquitin transferase BRCA1; RNF53; truncated BRCA1; truncated breast and ovarian cancer susceptibility protein 1; truncated breast and ovarian cancer sususceptibility protein 1; truncated breast cancer type 1 susceptibility protein | |
| Human recombinant protein fragment corresponding to amino acids 161-398 of human DAP3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction