missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA2 Antibody (BRCA2/2158), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24027G
This item is not returnable.
View return policy
Description
BRCA2 Monoclonal antibody specifically detects BRCA2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BRCA2 | |
| Monoclonal | |
| DyLight 488 | |
| BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2 | |
| Recombinant fragment (around aa 200-500) of human BRCA2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 675 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| BRCA2/2158 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction