missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCAA1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26619FR
This item is not returnable.
View return policy
Description
BRCAA1 Polyclonal antibody specifically detects BRCAA1 in Human samples. It is validated for Western Blot
Specifications
| BRCAA1 | |
| Polyclonal | |
| Western Blot | |
| 180 kDa Sin3-associated polypeptide, ARID domain-containing protein 4B, AT rich interactive domain 4B (RBP1- like), AT rich interactive domain 4B (RBP1-like), AT-rich interactive domain-containing protein 4B, BRCAA1DKFZp313M2420, breast cancer-associated antigen 1, Breast cancer-associated antigen BRCAA1, breast carcinoma-associated antigen, Histone deacetylase complex subunit SAP180, Rb-binding protein homolog, RBBP1L1RBP1L1BCAA, RBP1-like protein, retinoblastoma binding protein 1-like 1, Retinoblastoma-binding protein 1-like 1, SAP180MGC163290, SIN3A-associated protein 180, Sin3-associated polypeptide p180 | |
| The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human AT rich interactive domain 4B using the numbering given in entry NP_057458.4 (GeneID 51742). | |
| 0.1 mL | |
| Cancer, DNA Repair | |
| 51742 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction