missing translation for 'onlineSavingsMsg'
Learn More

BRD2 Antibody (7C1B10) - BSA Free, Novus Biologicals™

Product Code. p-200080599 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18239514 0.1 mL 0.1mL
18644727 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18239514 Supplier Novus Biologicals Supplier No. NBP261716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BRD2 Monoclonal specifically detects BRD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 7C1B10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3
Gene Symbols BRD2
Host Species Mouse
Immunogen Purified recombinant fragment of human BRD2 (AA: 227-364) expressed in E. Coli.
Molecular Weight of Antigen 45.5 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cellular Markers, DNA Repair, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6046
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.