missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD2, Mouse anti-Human, Clone: 3D10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006046-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: QTLEKIFLQKVASMPQEEQELVVTIPKNSHKKGAKLAALQGSVTSAHQVPAVSSVSHTALYTPPPEIPTTVLNIPHPSVISSPLLKSLHSSpecifications
| BRD2 | |
| Monoclonal | |
| Unconjugated | |
| bromodomain containing 2 | |
| D6S113E/DKFZp686N0336/FLJ31942/FSH/FSRG1/KIAA9001/NAT/RING3/RNF3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3D10 | |
| In 1x PBS, pH 7.4 | |
| NM_005104 | |
| BRD2 | |
| BRD2 (NP_005095, 167 a.a. ∼ 256 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6046 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction