missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BRD7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29305
This item is not returnable.
View return policy
Description
BRD7 Polyclonal antibody specifically detects BRD7 in Human samples. It is validated for Immunohistochemistry
Specifications
| BRD7 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| 75 kDa bromodomain protein, BP75bromodomain-containing 7, bromodomain containing 7, bromodomain-containing protein 7, CELTIX1NAG4, Protein CELTIX-1 | |
| Between 601 and 651 | |
| 25 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 29117 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction