missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD8 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79780JF646
This item is not returnable.
View return policy
Description
BRD8 Polyclonal antibody specifically detects BRD8 in Human samples. It is validated for Western Blot
Specifications
| BRD8 | |
| Polyclonal | |
| Western Blot | |
| bromodomain containing 8, bromodomain-containing protein 8, Skeletal muscle abundant protein, SMAP2, SMAPp120Skeletal muscle abundant protein 2, Thyroid hormone receptor coactivating protein of 120 kDa, trCP120 | |
| The immunogen recognized by this antibody maps to a region between residues 25 and 75 of human Bromodomain containing 8 using the numbering given in TrEMBL entry Q969M9 (GeneID 10902). | |
| 0.1 mL | |
| Cellular Markers | |
| 10902 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction