missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRD8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA530413
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (88%), Chicken (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
Especificaciones
| BRD8 | |
| Polyclonal | |
| Unconjugated | |
| BRD8 | |
| 2610007E11Rik; 4432404P07Rik; BRD8; bromodomain containing 8; bromodomain-containing protein 8; LOW QUALITY PROTEIN: bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; SMAP; SMAP2; Thyroid hormone receptor coactivating protein of 120 kDa; trCP120 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10902 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H0E9 | |
| BRD8 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human BRD8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido