missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82256-25ul
This item is not returnable.
View return policy
Description
BRD9 Polyclonal antibody specifically detects BRD9 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| BRD9 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| bromodomain containing 9, bromodomain-containing protein 9, DKFZp434D0711, DKFZp686L0539, FLJ13441, FLJ43530, LAVS3040, PRO9856, Rhabdomyosarcoma antigen MU-RMS-40.8, sarcoma antigen NY-SAR-29 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MGYLKRNGDGSLLYSVVNTAEPDADEEETHPVDLSSLSSKLLPGFTTLGFKDERRNKVTFLSSATTALSMQNNSVFGDLKSDEMELLYS | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 65980 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction