missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BrdU Monoclonal Antibody (MoBU-1), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112686
This item is not returnable.
View return policy
Description
BrdU Monoclonal Antibody for IHC, Flow
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
Specifications
| BrdU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| MoBU-1 | |
| FITC | |
| 5-Bromo-2-deoxyuridine; Bromodeoxyuridine | |
| Bromodeoxyuridine conjugated to Helix Pomatia Haemocyanin. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction