missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrdU Mouse, Clone: 85-2C8, Abnova™
Mouse monoclonal antibody raised against Bromodeoxyuridine (BrdU).
Brand: Abnova MAB13235.100ug
This item is not returnable.
View return policy
Specifications
| BrdU | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| Store at 4°C. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 85-2C8 | |
| Mouse monoclonal antibody raised against Bromodeoxyuridine (BrdU). | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| Bromodeoxyuridine (BrdU) conjugated with hemocyanine. | |
| 100 μg | |
| Primary | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction