missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry APC
Click to view available options
Quantity:
100 tests
Unit Size:
100 tests
Description
Includes
- BrdU
- DNase I
- Anti-BrdU APC Antibody (clone BU20A)
- BrdU Staining Buffer Concentrate (4X)
- Fixation/Permeabilization Diluent
- Cycling cells are incubated with 5-bromo-2′-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
- Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Application) | BrdU Staining |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Staining Kit |
| Format | Kit |
| Product Line | eBioscience |
| Quantity | 100 tests |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction