Learn More
BRE Mouse, anti-Human, CoraLite 594, Clone: 2E5A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-66356-20UL
 View more versions of this product
Product Code. 18034995
  • € 140.00 / 20µL
Description
The BRE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BRE. This antibody recognizes human, rat, mouse antigen. The BRE antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
BRE
2E5A5
CoraLite 594
Mouse
Primary
Human, Rat, Mouse
155 kDa
IgG2a
Immunofluorescence
1000 μg/mL
BRCA1 A complex subunit BRE, BRCC4, BRCC45, BRE
20 μL
9577
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.