missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT12 Antibody (OTI2G2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73106DL594
This item is not returnable.
View return policy
Description
NUDT12 Monoclonal antibody specifically detects NUDT12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NUDT12 | |
| Monoclonal | |
| DyLight 594 | |
| DKFZp761I172, EC 3.6.1.22, nucleoside diphosphate linked moiety X-type motif 12, Nucleoside diphosphate-linked moiety X motif 12, nudix (nucleoside diphosphate linked moiety X)-type motif 12, Nudix motif 12, peroxisomal NADH pyrophosphatase NUDT12 | |
| Recombinant protein fragment corresponding to amino acids 1-251 of human NUDT12 (NP_113626) produced in E.coli. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 83594 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2G2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction