missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brefeldin A, natural

Product Code. 10657631 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 10657631

Brand: Thermo Scientific Chemicals 297140250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4
Molecular Weight (g/mol) 280.36
MDL Number MFCD00083258,MFCD12913297
InChI Key KQNZDYYTLMIZCT-PNFJWZTBSA-N
Synonym brefeldin a, cyanein, decumbin, bfa
PubChem CID 133640201
IUPAC Name (1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one
SMILES C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

Specifications

Melting Point 201.0°C to 205.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 mg
Merck Index 15, 1374
Specific Rotation + 93.00
Solubility Information Solubility in water: .. Other solubilities: 10 mg/mL methanol
Formula Weight 280.35
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material Brefeldin A
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.