Learn More
Tocris Bioscience™ Brequinar sodium
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Brequinar sodium is a potent and selective dihydroorotate dehydrogenase (DHODH) inhibitor (IC50 = ∽20 nM). Causes depletion of pyrimidine nucleotides. Exhibits minimal inhibition against a panel of >400 kinases. Triggers differentiation of AML cell lines (ED50 = ∽ 1 μM. Note uridine concentration effects ED50 value). Causes differentiation and depletion of leukemia-initiating cells in vivo. Antimetabolite.
Specifications
Specifications
| Chemical Name or Material | 6-Fluoro-2-(2′-fluoro[1,1′-biphenyl]-4-yl)-3-methyl-4-quinolinecarboxylic acid sodium |
| CAS | 96201-88-6 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Target | DHODH Inhibitors |
| Molecular Formula | C23H14F2NO2Na |
| Formula Weight | Observed MW: 397.35 |