missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brevianamide F, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Brevianamide F |
| CAS | 38136-70-8 |
| Molecular Formula | C16H17N3O2 |
| Quantity | 5 mg |
| Synonym | (3S,8aS)-Hexahydro-3-(1H-indol-3-ylmethyl)pyrrolo[1,2-a]pyrazine-1,4-dione, Cyclo(L-Pro-L-Trp), Cyclo-L-tryptophyl-L-proline, L-Prolyl-L-tryptophan Anhydride Prolyltryptophanyldiketopiperazine, Cyclo-L-Prolyl-L-tryptophanyl |
| InChI Formula | InChI=1S/C16H17N3O2/c20-15-14-6-3-7-19(14)16(21)13(18-15)8-10-9-17-12-5-2-1-4-11(10)12/h1-2,4-5,9,13-14,17H,3,6-8H2,(H,18,20)/t13-,14-/m0/s1 |
| SMILES | O=C1N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N4CCC[C@@H]14 |
| IUPAC Name | (3S,8aS)-3-(1H-indol-3-ylmethyl)-2,3,6,7,8,8a-hexahydropyrrolo[1,2-a]pyrazine-1,4-dione |
| Molecular Weight (g/mol) | 283.33 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction