missing translation for 'onlineSavingsMsg'
Learn More

Brevianamide F, TRC

Product Code. 30341691
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30341691

Brand: Toronto Research Chemicals TRCB67739550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brevianamide F
CAS 38136-70-8
Molecular Formula C16H17N3O2
Quantity 50 mg
Synonym (3S,8aS)-Hexahydro-3-(1H-indol-3-ylmethyl)pyrrolo[1,2-a]pyrazine-1,4-dione, Cyclo(L-Pro-L-Trp), Cyclo-L-tryptophyl-L-proline, L-Prolyl-L-tryptophan Anhydride Prolyltryptophanyldiketopiperazine, Cyclo-L-Prolyl-L-tryptophanyl
InChI Formula InChI=1S/C16H17N3O2/c20-15-14-6-3-7-19(14)16(21)13(18-15)8-10-9-17-12-5-2-1-4-11(10)12/h1-2,4-5,9,13-14,17H,3,6-8H2,(H,18,20)/t13-,14-/m0/s1
SMILES O=C1N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N4CCC[C@@H]14
IUPAC Name (3S,8aS)-3-(1H-indol-3-ylmethyl)-2,3,6,7,8,8a-hexahydropyrrolo[1,2-a]pyrazine-1,4-dione
Molecular Weight (g/mol) 283.33
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.