missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brg1 Antibody (BRG1/6531R), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20937JF549
This item is not returnable.
View return policy
Description
Brg1 Monoclonal antibody specifically detects Brg1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| Brg1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| ATP-dependent helicase SMARCA4, BAF190, BAF190A, brahma protein-like 1, BRG1-associated factor 190A, BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4, BRM/SWI2-related gene 1, EC 3.6.1, EC 3.6.4.-, FLJ39786, global transcription activator homologous sequence, homeotic gene regulator, hSNF2b, Mitotic growth and transcription activator, nuclear protein GRB1, Protein brahma homolog 1, Protein BRG-1, RTPS2, SNF2, SNF2B, SNF2-BETA, SNF2L4, SNF2LB, SNF2-like 4, subfamily a, member 4, sucrose nonfermenting-like 4, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI2, transcription activator BRG1 | |
| Recombinant fragment (around aa 200-400) of human Brg1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Chromatin Modifiers, Neuroscience, Stem Cells | |
| 6597 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| BRG1/6531R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction