Learn More
Brg1 Antibody (CL13779) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24575AF750
Product Code. 30296271
  • € 737.00 / 0.10mL
Description
Brg1 Monoclonal antibody specifically detects Brg1 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Brg1
Monoclonal
Alexa Fluor 750
ATP-dependent helicase SMARCA4, BAF190, BAF190A, brahma protein-like 1, BRG1-associated factor 190A, BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4, BRM/SWI2-related gene 1, EC 3.6.1, EC 3.6.4.-, FLJ39786, global transcription activator homologous sequence, homeotic gene regulator, hSNF2b, Mitotic growth and transcription activator, nuclear protein GRB1, Protein brahma homolog 1, Protein BRG-1, RTPS2, SNF2, SNF2B, SNF2-BETA, SNF2L4, SNF2LB, SNF2-like 4, subfamily a, member 4, sucrose nonfermenting-like 4, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI2, transcription activator BRG1
This antibody was developed using a synthetic peptide derived from P51532, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Chromatin Modifiers, Neuroscience, Stem Cells
6597
Store at 4⪚C in the dark.
IgG1
Western Blot, Immunocytochemistry/Immunofluorescence
CL13779
50mM Sodium Borate
Mouse
Protein A purified