missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRICD5 Polyclonal Antibody

BRICD5 Polyclonal Antibody
Brand: Invitrogen PA553951
This item is not returnable.
View return policy
Description
BRICD5 Polyclonal Antibody for Western Blot, IHC
Specifications
| BRICD5 | |
| Polyclonal | |
| Unconjugated | |
| BRICD5 | |
| BRICD5; BRICHOS domain containing 5; BRICHOS domain-containing protein 5; BRICHOS domain-containing protein C16orf79; C16orf79 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283870 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PL45 | |
| BRICD5 | |
| Recombinant protein corresponding to Human BRICD5. Recombinant protein control fragment (Product #RP-91049). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction