missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Brilliant Green
Click to view available options
Quantity:
25 g
100 g
Description
Brilliant Green a stain used in bacteriological media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Brilliant Green a stain used in bacteriological media.
Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).
Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
Brilliant Green a stain used in bacteriological media.
Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).
Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | 210°C (decomposition) |
Beilstein | 3901207 |
Merck Index | 14,1374 |
Solubility Information | Soluble in water (100mg/ml at 25°C),ethanol (50mg/ml),methanol,and 2-methoxyethanol (60mg/ml). |
Formula Weight | 482.65 |
Quantity | 25 g |
Chemical Name or Material | Brilliant Green |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'